Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300001160.mm.1 |
NM_001039090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001160.mm.1 |
NM_001271772.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001160.mm.1 |
NM_011386.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001160.mm.1 |
TC0300000210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001162.mm.1 |
NM_001039090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001162.mm.1 |
NM_001271772.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001162.mm.1 |
NM_011386.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001162.mm.1 |
TC0300000210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001163.mm.1 |
NM_001039090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001163.mm.1 |
NM_001271772.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001163.mm.1 |
NM_011386.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001163.mm.1 |
TC0300000210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001161.mm.1 |
NM_001039090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001161.mm.1 |
NM_001271772.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001161.mm.1 |
NM_011386.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001161.mm.1 |
TC0300000210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001166.mm.1 |
NM_001039090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001166.mm.1 |
NM_001271772.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001166.mm.1 |
NM_011386.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001166.mm.1 |
TC0300000210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001168.mm.1 |
NM_001039090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001168.mm.1 |
NM_001271772.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001168.mm.1 |
NM_011386.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001168.mm.1 |
TC0300000210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001165.mm.1 |
NM_001039090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001165.mm.1 |
NM_011386.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001165.mm.1 |
TC0300000210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001167.mm.1 |
NM_001039090.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001167.mm.1 |
NM_011386.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001167.mm.1 |
TC0300000210.mm.1 |
MTA TC |