Assay Details
| Gene Symbol: Timm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013896.3 | NP_038924.1 | 1 - 2 | 117 | 78 | |
| GenBank mRNA | AI119857.1 | - | 1 - 2 | 98 | 78 | |
| BC024370.1 | - | 1 - 2 | 90 | 4976059 | 78 | |
| CO809703.1 | - | 48 | 78 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC024370.1, NM_013896.3 transcripts.
Set Membership: |
|

Timm9
Genomic Map