Assay Details
| Gene Symbol: Dedd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128609.1 | NP_001122081.1 | 1 - 2 | 387 | 93 | |
| GenBank mRNA | AK157609.1 | - | 1 - 2 | 333 | 93 | |
| BM947911.1 | - | 1 - 2 | 387 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001128609.1 transcripts.
Set Membership: |
|

Dedd
Genomic Map