Assay Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025505.4 | NP_079781.2 | 4 - 5 | 349 | 72 | |
| GenBank mRNA | AK029787.1 | - | 4 - 5 | 350 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Blzf1| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025505.4 | NP_079781.2 | 4 - 5 | 349 | 72 | |
| GenBank mRNA | AK029787.1 | - | 4 - 5 | 350 | 72 | |
Set Membership: |
|