Assay Details
| Gene Symbol: Slc26a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290320.1 | NP_001277249.1 | 7 - 8 | 1223 | 73 | |
| GenBank mRNA | BC027076.1 | - | 7 - 8 | 1223 | 5361175 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a8
Genomic Map