Assay Details
| Gene Symbol: Alkbh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143105.1 | - | 1 - 2 | 200 | 87 | |
| BC050856.1 | - | 1 - 2 | 189 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alkbh1| Gene Symbol: Alkbh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK143105.1 | - | 1 - 2 | 200 | 87 | |
| BC050856.1 | - | 1 - 2 | 189 | 87 | ||
Set Membership: |
|