Assay Details
| Gene Symbol: Thsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172485.3 | NP_766073.2 | 19 - 20 | 4015 | 77 | |
| GenBank mRNA | AK166575.1 | - | 7 - 8 | 1199 | 77 | |
| BC063250.1 | - | 19 - 20 | 3845 | 6410178 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Thsd7b
Genomic Map