Assay Details
| Gene Symbol: Smarcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031842.2 | NP_114030.2 | 10 - 11 | 1306 | 62 | |
| GenBank mRNA | AK075611.1 | - | 6 - 7 | 643 | 62 | |
| BC026783.1 | - | 10 - 11 | 1218 | 4481097 | 62 | |
| BC059921.1 | - | 10 - 11 | 1276 | 62 | ||
| BC079839.1 | - | 10 - 11 | 1252 | 6810462 | 62 | |
| M25773.1 | - | 10 - 11 | 1262 | 62 | ||
| U66620.1 | - | 10 - 11 | 1262 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smarcd1
Genomic Map