Assay Details
| Gene Symbol: Epx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007946.2 | NP_031972.2 | 11 - 12 | 2078 | 74 | |
| GenBank mRNA | AK132416.1 | - | 6 - 7 | 1203 | 74 | |
| BC052881.1 | - | 11 - 12 | 2060 | 74 | ||
| D78353.1 | - | 11 - 12 | 2084 | 74 | ||
| L77979.1 | - | 11 - 12 | 2062 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Epx
Genomic Map