Assay Details
| Gene Symbol: Epx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007946.2 | NP_031972.2 | 4 - 5 | 591 | 146 | |
| GenBank mRNA | BC052881.1 | - | 4 - 5 | 573 | 146 | |
| D78353.1 | - | 4 - 5 | 597 | 146 | ||
| L77979.1 | - | 4 - 5 | 575 | 146 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Epx
Genomic Map