Assay Details
| Gene Symbol: Epx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007946.2 | NP_031972.2 | 6 - 7 | 927 | 94 | |
| GenBank mRNA | BC052881.1 | - | 6 - 7 | 94 | ||
| D78353.1 | - | 6 - 7 | 933 | 94 | ||
| L77979.1 | - | 6 - 7 | 911 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epx
Genomic Map