Assay Details
| Gene Symbol: Ngfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033217.3 | NP_150086.2 | 1 - 2 | 219 | 95 | |
| GenBank mRNA | AF105292.1 | - | 1 - 2 | 77 | 95 | |
| AK037248.1 | - | 1 - 2 | 220 | 95 | ||
| BB630380.1 | - | 1 - 2 | 220 | 95 | ||
| BC038365.1 | - | 1 - 2 | 210 | 5367638 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF105292.1, AK037248.1 transcripts.
Set Membership: |
|

Ngfr
Genomic Map