Assay Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080844.4 | NP_543120.1 | 4 - 5 | 840 | 104 | |
| GenBank mRNA | AK050295.1 | - | 3 - 4 | 682 | 104 | |
| BC019447.1 | - | 4 - 5 | 800 | 5125534 | 104 | |
| BC033377.1 | - | 3 - 4 | 684 | 5123932 | 104 | |
| S47225.1 | - | 3 - 4 | 643 | 104 | ||
More Information
Set Membership: |
|

Serpinc1
Genomic Map