Assay Details
| Gene Symbol: Agpat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026644.2 | NP_080920.2 | 6 - 7 | 979 | 81 | |
| GenBank mRNA | AK005139.1 | - | 6 - 7 | 980 | 81 | |
| AK166917.1 | - | 6 - 7 | 933 | 81 | ||
| BC031179.1 | - | 6 - 7 | 1102 | 81 | ||
| BC047281.1 | - | 6 - 7 | 910 | 6477257 | 81 | |
More Information
Set Membership: |
|

Agpat4
Genomic Map