Assay Details
| Gene Symbol: Wdr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145514.5 | NP_663489.4 | 6 - 7 | 1504 | 144 | |
| GenBank mRNA | AK075743.1 | - | 6 - 7 | 896 | 144 | |
| AK082326.1 | - | 7 - 8 | 799 | 144 | ||
| BC020044.1 | - | 2 - 3 | 249 | 144 | ||
| BC058601.1 | - | 4 - 5 | 500 | 144 | ||
| BC094233.1 | - | 3 - 4 | 389 | 144 | ||
| BC139336.1 | - | 6 - 7 | 666 | 8862358 | 144 | |
| BC139337.1 | - | 6 - 7 | 666 | 8862359 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr26
Genomic Map