| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400015750.mm.1 |
NM_080728.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015750.mm.1 |
NM_080728.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015750.mm.1 |
TC1400002075.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400015751.mm.1 |
NM_080728.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015751.mm.1 |
NM_080728.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015751.mm.1 |
TC1400002075.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400015752.mm.1 |
NM_080728.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015752.mm.1 |
NM_080728.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015752.mm.1 |
TC1400002075.mm.1 |
MTA TC |