Assay Details
| Gene Symbol: Gm31739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076901.1 | - | 2 - 3 | 682 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prostaglandin F receptor | Ptgfr |
AI957154, PGF, fp |
-152.3kb | Anti-sense | Chr. 3 - 151798610 - 151837528 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 31881 |
Gm31881 | 91.7kb | Sense | Chr. 3 - 152084026 - 152107379 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31739
Genomic Map