Assay Details
| Gene Symbol: Fbxw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013908.4 | NP_038936.1 | 6 - 7 | 1236 | 60 | |
| GenBank mRNA | AF176520.1 | - | 6 - 7 | 1197 | 60 | |
| AK019715.2 | - | 6 - 7 | 1237 | 60 | ||
| AK154911.1 | - | 5 - 6 | 2175 | 60 | ||
| AK187599.1 | - | 1 - 2 | 199 | 60 | ||
| BC010776.1 | - | 6 - 7 | 1163 | 4211592 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxw5
Genomic Map