Assay Details
| Gene Symbol: Cry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009963.4 | NP_034093.1 | 4 - 5 | 689 | 77 | |
| GenBank mRNA | AF156987.1 | - | 4 - 5 | 657 | 77 | |
| AK041696.1 | - | 4 - 5 | 657 | 77 | ||
| AK133781.1 | - | 4 - 5 | 664 | 77 | ||
| AK172994.1 | - | 4 - 5 | 621 | 77 | ||
| BC054794.1 | - | 4 - 5 | 645 | 6834104 | 77 | |
| BC066799.1 | - | 4 - 5 | 648 | 30103169 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cry2
Genomic Map