Assay Details
| Gene Symbol: Slc43a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021398.3 | NP_067373.2 | 8 - 9 | 1135 | 128 | |
| GenBank mRNA | BC089603.1 | - | 7 - 8 | 835 | 6758493 | 128 |
| BU516072.1 | - | 2 - 3 | 171 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc43a3
Genomic Map