| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200001398.mm.1 |
NM_001001334.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001398.mm.1 |
NM_001001334.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001398.mm.1 |
TC0200000303.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200001393.mm.1 |
NM_001001334.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001393.mm.1 |
NM_001001334.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001393.mm.1 |
TC0200000303.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200001397.mm.1 |
NM_001001334.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001397.mm.1 |
NM_001001334.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001397.mm.1 |
TC0200000303.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200001396.mm.1 |
NM_001001334.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001396.mm.1 |
NM_001001334.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001396.mm.1 |
TC0200000303.mm.1 |
MTA TC |