Assay Details
| Gene Symbol: Ccdc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138603.3 | NP_613069.3 | 3 - 4 | 484 | 67 | |
| GenBank mRNA | AF229637.1 | - | 3 - 4 | 425 | 67 | |
| AK010536.1 | - | 3 - 4 | 402 | 67 | ||
| AK039573.1 | - | 3 - 4 | 67 | |||
| AK209776.1 | - | 2 - 3 | 162 | 67 | ||
| BC011195.1 | - | 3 - 4 | 389 | 4188459 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc22
Genomic Map