Assay Details
| Gene Symbol: Scn10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205321.1 | NP_001192250.1 | 12 - 13 | 1821 | 84 | |
| NM_009134.3 | NP_033160.2 | 12 - 13 | 1821 | 84 | ||
| GenBank mRNA | AK135787.1 | - | 9 - 10 | 1442 | 84 | |
| AK157866.1 | - | 12 - 13 | 1821 | 84 | ||
| AY538273.1 | - | 11 - 12 | 1751 | 84 | ||
More Information
Set Membership: |
|

Scn10a
Genomic Map