Assay Details
Gene Symbol: Adcy5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012765.4 | NP_001012783.3 | 19 - 20 | 3997 | 68 | |
GenBank mRNA | AK147590.1 | - | 4100 | 68 | ||
AK147649.1 | - | 4757 | 68 | |||
AK160942.1 | - | 19 - 20 | 3997 | 68 | ||
BC035550.1 | - | 2135 | 68 | |||
BC090846.1 | - | 19 - 20 | 3732 | 30363147 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
