Assay Details
| Gene Symbol: Krtap6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010673.2 | NP_034803.2 | 1 | 738 | 81 | |
| GenBank mRNA | AA763314.1 | - | 1 | 233 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985648 | NM_010673.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985648 | AA763314.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985648 | NM_010673.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985648 | AA763314.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30950943 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08677739 | - | - |
| Whole Mouse Genome G4122A | A_51_P350976 | D89902.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114365 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08677739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30950903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12641590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12764825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12641567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12686688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12641587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12519076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12951054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12641569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30950920 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10440725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440726 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449919_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10440724 | - | - |
| Mouse Genome 430 2.0 Array | 1449919_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449919_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16701_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16701_PM_at | - | - |
| Mouse Expression Set 430 | 1449919_at | - | - |
| Mouse Expression Set 430 | 1449919_PM_at | - | - |
| Murine Genome U74v2 Set | 92253_PM_at | - | - |
| Murine Genome U74v2 Set | 92253_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449919_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449919_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449919_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449919_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 130064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000130064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2761643 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2761643 | - | - |
| mouse-6 v1.1 (Illumina) | scl0016701.1_246-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 130064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0016701.1_246-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 130064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2795861 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2761643 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2795861 | - | - |

Krtap6-2
Genomic Map