Assay Details
| Gene Symbol: Prodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011172.2 | NP_035302.2 | 7 - 8 | 960 | 74 | |
| GenBank mRNA | AF120279.1 | - | 7 - 8 | 917 | 74 | |
| BC125327.1 | - | 7 - 8 | 763 | 40129842 | 74 | |
| U80020.1 | - | 7 - 8 | 909 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prodh
Genomic Map