Assay Details
| Gene Symbol: Atf7ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153123.2 | NP_694763.2 | 11 | 2527 | 106 | |
| GenBank mRNA | AK145634.1 | - | 11 | 2485 | 106 | |
| BC018510.1 | - | 1 | 975 | 106 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atf7ip2
Genomic Map