Assay Details
| Gene Symbol: Zfp952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045559.1 | NP_001039024.1 | 3 - 4 | 308 | 138 | |
| GenBank mRNA | BC051011.1 | - | 3 - 4 | 250 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008186 | BC051011.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008186 | BC057593.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008186 | NM_001045559.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674592 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578001 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967394 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1485 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122166 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107564 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008186 | BC051011.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008186 | BC057593.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008186 | NM_001045559.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P391058 | BC051011.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P391058 | BC057593.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P391058 | NM_001045559.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13168883 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13168878 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13168896 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13168893 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13168901 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13168891 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13168886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31148023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13221133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13365235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13221131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13168888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13168900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13453451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13465594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13168889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13168893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147999 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10443925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443920 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457677_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10443918 | - | - |
| Mouse Genome 430 2.0 Array | 1457677_at | - | - |
| Mouse Genome 430 2.0 Array | 1457677_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240067_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240067_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700005211.mm.1 | NM_001045559.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005211.mm.1 | NM_001045559.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005211.mm.1 | TC1700000572.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005202.mm.1 | NM_001045559.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005202.mm.1 | NM_001045559.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005202.mm.1 | TC1700000572.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005204.mm.1 | NM_001045559.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005204.mm.1 | NM_001045559.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005204.mm.1 | TC1700000572.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005207.mm.1 | NM_001045559.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005207.mm.1 | NM_001045559.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005207.mm.1 | TC1700000572.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005208.mm.1 | NM_001045559.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005208.mm.1 | NM_001045559.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005208.mm.1 | TC1700000572.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005210.mm.1 | NM_001045559.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005210.mm.1 | NM_001045559.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005210.mm.1 | TC1700000572.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1457677_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457677_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 708480 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2651505 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2651505 | - | - |

Zfp952
Genomic Map