Assay Details
| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027492.1 | 4 - 5 | 801 | 94 | ||
| GenBank mRNA | AK030553.1 | - | 4 - 5 | 774 | 94 | |
| BC094508.1 | - | 4 - 5 | 744 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspa13
Genomic Map