Assay Details
| Gene Symbol: Rd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC017602.1 | - | 2 - 3 | 570 | 4503004 | 64 |
| BC089516.1 | - | 2 - 3 | 780 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rd3
Genomic Map