| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500028265.mm.1 |
TC0500003180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500028265.mm.1 |
NM_011852.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500028263.mm.1 |
TC0500003180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500028257.mm.1 |
TC0500003180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500028257.mm.1 |
NM_011852.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500028260.mm.1 |
TC0500003180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500028260.mm.1 |
NM_011852.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500028261.mm.1 |
TC0500003180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500028261.mm.1 |
NM_011852.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500028266.mm.1 |
TC0500003180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500028266.mm.1 |
NM_011852.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500028258.mm.1 |
TC0500003180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500028258.mm.1 |
NM_011852.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500028264.mm.1 |
TC0500003180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500028264.mm.1 |
NM_011852.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500028259.mm.1 |
TC0500003180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500028259.mm.1 |
NM_011852.3 |
Refseq |