Assay Details
| Gene Symbol: 1700025O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006362.1 | - | 2 | 374 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P399563 | AK006362.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P399563 | AK006362.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P399563 | AK006362.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P399563 | AK006362.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03497240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03887987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03831438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03259311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03739489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22779994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22780030 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 452470 | - | - |

1700025O08Rik
Genomic Map