Assay Details
| Gene Symbol: 1700034B16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006590.1 | - | 2 | 668 | 129 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CBP80/20-dependent translation initiation factor | Ctif | Gm672 | -84.3kb | Anti-sense | Chr. 18 - 75431205 - 75697696 on NCBI Build GRCm38 |
| Downstream | zinc finger and BTB domain containing 7C | Zbtb7c |
B230208J24Rik, Kr-pok, Zbtb36 |
37.9kb | Sense | Chr. 18 - 75820178 - 76148564 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700034B16Rik
Genomic Map