Assay Details
| Gene Symbol: Ucn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031250.5 | NP_112540.2 | 2 | 740 | 82 | |
| GenBank mRNA | AK148451.1 | - | 2 | 741 | 82 | |
| BC119433.1 | - | 1 | 225 | 40041175 | 82 | |
| BC119434.1 | - | 1 | 222 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ucn3
Genomic Map