Assay Details
| Gene Symbol: 1700013K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005961.1 | - | 1 | 50 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A730043L09 gene | A730043L09Rik | A730043L09 | -23.3kb | Sense | Chr. 9 - 62242596 - 62244100 on NCBI Build GRCm38 |
| Downstream | sperm equatorial segment protein 1 | Spesp1 |
4921508E09Rik, AI428149 |
2.9kb | Sense | Chr. 9 - 62270729 - 62282179 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700013K18Rik
Genomic Map