Assay Details
Gene Symbol: 4930509B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015731.1 | - | 1 | 754 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA D930015E06 gene | D930015E06Rik |
Kiaa0922, mKIAA0922 |
Anti-sense | Chr. 3 - 83898205 - 84040182 on NCBI Build GRCm38 | |
Downstream | meiotic nuclear divisions 1 homolog (S. cerevisiae) | Mnd1 |
2610034E18Rik, AI591601, Gaj |
58.7kb | Anti-sense | Chr. 3 - 84087933 - 84156861 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
