Assay Details
| Gene Symbol: 4930568B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016246.1 | - | 1 | 751 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | brain protein I3 | Bri3 | I3 | -98.1kb | Anti-sense | Chr. 5 - 144244437 - 144446757 on NCBI Build GRCm38 |
| Downstream | neuronal pentraxin 2 | Nptx2 |
Narp, np2 |
129.0 | Anti-sense | Chr. 5 - 144545887 - 144557478 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930568B11Rik
Genomic Map