Assay Details
| Gene Symbol: Gm20556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040347.1 | 2 - 3 | 731 | 108 | ||
| GenBank mRNA | AK079813.1 | - | 2 - 3 | 732 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | proline rich 5 (renal) | Prr5 |
AU043908, Arhgap8, C030017C09Rik, C78947, Protor-1, Protor1 |
-8.2kb | Anti-sense | Chr. 15 - 84663171 - 84706409 on NCBI Build GRCm38 |
| Downstream | Rho GTPase activating protein 8 | Arhgap8 | 3110043J09Rik | 906.0 | Anti-sense | Chr. 15 - 84720052 - 84772857 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm20556
Genomic Map