Assay Details
| Gene Symbol: 5730408K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011444.1 | - | 1 - 2 | 393 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LBH domain containing 1 | Lbhd1 | Gm21743 | Anti-sense | Chr. 19 - 8883878 - 8890759 on NCBI Build GRCm38 | |
| Downstream | integrator complex subunit 5 | Ints5 |
1110055N21Rik, mKIAA1698 |
3.8kb | Anti-sense | Chr. 19 - 8892987 - 8897890 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

5730408K05Rik
Genomic Map