Assay Details
| Gene Symbol: A930009E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020797.1 | - | 1 | 251 | 104 | |
| AK020836.1 | - | 1 | 282 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier organic anion transporter family, member 6c1 |
Slco6c1 |
4933404A18Rik, C79179 |
-509.1kb | Sense | Chr. 1 - 97059447 - 97128340 on NCBI Build GRCm38 |
| Downstream |
DNA segment, Chr 1, ERATO Doi 622, expressed |
D1Ertd622e |
AI987691, AW047450 |
5.6kb | Sense | Chr. 1 - 97643902 - 97662018 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A930009E08Rik
Genomic Map