Assay Details
| Gene Symbol: Rpl39l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026594.2 | NP_080870.1 | 1 - 2 | 100 | 103 | |
| GenBank mRNA | AK005645.1 | - | 1 - 2 | 90 | 103 | |
| AK019681.1 | - | 1 - 2 | 102 | 103 | ||
| BC023885.1 | - | 1 - 2 | 90 | 5151514 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005645.1, AK019681.1, BC023885.1, NM_026594.2 transcripts.
Set Membership: |
|

Rpl39l
Genomic Map