Assay Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026883.3 | NP_081159.1 | 2 - 3 | 144 | 82 | |
| GenBank mRNA | AK005211.1 | - | 2 - 3 | 536 | 82 | |
| AK150218.1 | - | 2 - 3 | 543 | 82 | ||
| AK193505.1 | - | 1 - 2 | 72 | 82 | ||
| BC019498.1 | - | 2 - 3 | 502 | 4460879 | 82 | |
| BC057658.1 | - | 2 - 3 | 183 | 3153717 | 82 | |
| BC061033.1 | - | 2 - 3 | 144 | 6590939 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam216a
Genomic Map