Assay Details
| Gene Symbol: Zfp955a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029952.3 | NP_084228.2 | 4 | 3444 | 103 | |
| GenBank mRNA | AK146709.1 | - | 4 | 3442 | 103 | |
| BC043067.1 | - | 4 | 3407 | 5709349 | 103 | |
| BC094586.1 | - | 4 | 3424 | 30620063 | 103 | |
| BG067234.2 | - | 1 | 103 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323658 | BC094586.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323658 | BG067234.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323658 | BC043067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323658 | AK146709.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323658 | NM_029952.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323658 | BC094586.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323658 | BG067234.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323658 | BC043067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323658 | AK146709.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P323658 | NM_029952.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416294 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331783 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525309 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 797326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846035 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513937 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308122 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851187 | AK045783.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851187 | BC080289.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851187 | NM_001003950.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506731 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89362 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 586925 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126568 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38154 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130419 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177991 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150460 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133605 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108250 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323658 | BC094586.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323658 | BG067234.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323658 | BC043067.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323658 | AK146709.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323658 | NM_029952.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323658 | BC094586.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323658 | BG067234.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323658 | BC043067.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323658 | AK146709.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P323658 | NM_029952.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P239023 | AK161843.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P239023 | NM_001142957.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P239023 | AK021244.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P239023 | BC094586.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P239023 | BC043067.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P239023 | AK161843.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P239023 | NM_001142957.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P239023 | AK021244.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P239023 | BC094586.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P239023 | BC043067.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P359473 | AK013605.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31148566 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03308864 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762763 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150460 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762614 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108250 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762714 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762665 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133605 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762837 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03308919 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118284 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762594 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762767 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762651 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762844 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762664 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762777 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03309005 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762599 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762863 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177991 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762781 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762840 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762729 | - | - |
| Whole Mouse Genome G4122A | A_51_P359473 | AK013605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P239023 | AK161843.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P239023 | NM_001142957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P239023 | AK021244.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P239023 | BC094586.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P239023 | BC043067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P239023 | AK161843.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P239023 | NM_001142957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P239023 | AK021244.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P239023 | BC094586.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P239023 | BC043067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413995 | AK032339.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323658 | BC094586.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323658 | BG067234.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323658 | BC043067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323658 | AK146709.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323658 | NM_029952.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323658 | BC094586.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323658 | BG067234.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323658 | BC043067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323658 | AK146709.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P323658 | NM_029952.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105453 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115839 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08762840 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03308864 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177991 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08762714 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133605 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03309005 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150460 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118284 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108250 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08762777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13563424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13455732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13320953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13455742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13455748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31148594 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10443942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443944 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443941 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434896_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10443940 | - | - |
| Mouse Genome 430 2.0 Array | 1453198_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441348_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441348_at | - | - |
| Mouse Genome 430 2.0 Array | 1434896_at | - | - |
| Mouse Genome 430 2.0 Array | 1434896_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429560_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453198_at | - | - |
| Mouse Genome 430 2.0 Array | 1429560_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700017246.mm.1 | NM_029952.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017246.mm.1 | NM_029952.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017246.mm.1 | TC1700001864.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1434896_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434896_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441348_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441348_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5720411 | - | - |
| MouseWG-6 v2 Expression BeadChip | 1580348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720411 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1580348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6660373 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660373 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720411 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001580348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215336 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235833 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2523128 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215336 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235833 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2523128 | - | - |
| mouse-6 v1.1 (Illumina) | scl0077652.1_81-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0077652.1_86-S | - | - |
| mouse-6 v1.1 (Illumina) | scl077652.1_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1580348 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720411 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660373 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0077652.1_86-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0077652.1_81-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl077652.1_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720411 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1580348 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660373 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2523128 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215336 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235833 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl077652.1_16-S | - | - |

Zfp955a
Genomic Map