Assay Details
| Gene Symbol: Gm7550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033689.1 | 1 - 2 | 139 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700104L18 gene | 1700104L18Rik | -145.4kb | Anti-sense | Chr. 12 - 54233615 - 54242320 on NCBI Build GRCm38 | |
| Downstream |
serine palmitoyltransferase, small subunit A |
Sptssa |
1110002B05Rik, AA407909, AA409588, AU041967, Ssspta |
254.1kb | Sense | Chr. 12 - 54645374 - 54656572 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm7550
Genomic Map