Assay Details
| Gene Symbol: Crlf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277106.1 | NP_001264035.1 | 5 - 6 | 911 | 82 | |
| NM_018776.2 | NP_061246.1 | 5 - 6 | 911 | 82 | ||
| GenBank mRNA | AF046060.1 | - | 5 - 6 | 886 | 82 | |
| AF120152.1 | - | 5 - 6 | 820 | 82 | ||
| AK037880.1 | - | 5 - 6 | 859 | 82 | ||
| AK078569.1 | - | 5 - 6 | 857 | 82 | ||
| AK137528.1 | - | 5 - 6 | 863 | 82 | ||
| AK146798.1 | - | 5 - 6 | 859 | 82 | ||
| AK195261.1 | - | 1 - 2 | 158 | 82 | ||
| AK197632.1 | - | 1 - 2 | 166 | 82 | ||
| BC004780.1 | - | 5 - 6 | 858 | 3257685 | 82 | |
More Information
Set Membership: |
|

Crlf3
Genomic Map