Assay Details
| Gene Symbol: Aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC057681.1 | - | 1 - 2 | 172 | 3499299 | 100 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC057681.1 transcripts.
Set Membership: |
|

Aebp2
Genomic Map