Assay Details
| Gene Symbol: Krt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010661.2 | NP_034791.2 | 7 - 8 | 1411 | 70 | |
| GenBank mRNA | AK053362.1 | - | 7 - 8 | 1412 | 70 | |
| AK143182.1 | - | 7 - 8 | 1410 | 70 | ||
| BC131911.1 | - | 7 - 8 | 1407 | 40130188 | 70 | |
| U02880.1 | - | 7 - 8 | 1408 | 70 | ||
More Information
Set Membership: |
|

Krt12
Genomic Map