Assay Details
| Gene Symbol: Prss27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175440.4 | NP_780649.1 | 3 - 4 | 308 | 72 | |
| GenBank mRNA | AF542056.1 | - | 3 - 4 | 244 | 72 | |
| AK080281.1 | - | 3 - 4 | 309 | 72 | ||
| BC117009.1 | - | 3 - 4 | 291 | 40126328 | 72 | |
| BC117011.1 | - | 3 - 4 | 291 | 40126330 | 72 | |
More Information
Set Membership: |
|

Prss27
Genomic Map