Assay Details
| Gene Symbol: Ccnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183015.3 | NP_898836.2 | 5 - 6 | 432 | 89 | |
| GenBank mRNA | AJ555464.1 | - | 5 - 6 | 433 | 89 | |
| AJ555465.1 | - | 5 - 6 | 417 | 89 | ||
| BC132411.1 | - | 4 - 5 | 386 | 40130688 | 89 | |
| BC138355.1 | - | 4 - 5 | 386 | 8861375 | 89 | |
More Information
Set Membership: |
|

Ccnb3
Genomic Map