Assay Details
| Gene Symbol: Sppl2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082535.1 | NP_001076004.1 | 1 | 1318 | 73 | |
| NM_199019.2 | NP_950184.2 | 1 | 1318 | 73 | ||
| GenBank mRNA | AK077122.1 | - | 1 | 1319 | 73 | |
| BC090973.1 | - | 1 | 1293 | 73 | ||
| BC100418.1 | - | 1 | 1291 | 6706317 | 73 | |
More Information
Set Membership: |
|

Sppl2c
Genomic Map